(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA3036-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+31.0 to +34.0 deg (C=1, 6N HCl)
Specification Test Results
About this item
:
:212.2026 g/mol
EINECS:221-001-4
:GHS07
CAS Number:2964-48-9
Formula:C₉H₁₂N₂O₄
MDL Number:MFCD00007359