AOL-biotin conjugate
1 mL
TCIAA2659-1ML
: TCI
- :Refrigerator
Documents
- Appearance:Colorless to Light yellow clear liquid
- Concentration (Lowry Methoed):0.9 to 1.5 mg/mL
- Molecular weight:30 to 40 kDa
- Suitability for use in ELISA tests:min. 1.0 (human Lactoferrin, 10 micro g/mL, OD450)
- Fucose inhibition test:to pass test
- Pk:1 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



