
4-(4-Aminophenyl)morpholin-3-one ≥98.0% (by GC, titration analysis)
: TCI
:438056-69-0
:
:C₁₀H₁₂N₂O₂
:MFCD08236742
:192.2145 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:171.0 to 175.0 deg-C
Frequently Bought Together