Azido-PEG4-NHS ester
TCIAA2388-25MG
: TCI
:
- Pk:25 mg
- Seq:2315
- Appearance:Colorless to Light orange to Yellow clear liquid
- Elemental analysis (Nitrogen):13.00 to 15.90 %
:
:388.38 g/mol
:Freezer
:944251-24-5
:C₁₅H₂₄N₄O₈
:MFCD13184948