Ammonium acetate ≥97.0% (by titrimetric analysis)
Catalog # TCIAA2269-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Appearance:White or Colorless powder to crystal
- Purity (Formol Titration):min. 97.0 %
Specification Test Results
About this item
:
:77.08 g/mol
Melting Pt:114°C
EINECS:211-162-9
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066