
2-[(Acetylthio)methyl]-3-phenylpropionic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:91702-98-6
:
Formula:C₁₂H₁₄O₃S
MDL Number:MFCD00236751
:238.30 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to lump
- Purity (GC):min. 93.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together