Order Entry
Denmark
ContactUsLinkComponent
(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene ≥95.0% (by GC)
Irritant
(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene ≥95.0% (by GC)
Catalog # TCIAA2128-5G
Supplier:  TCI
CAS Number:  
Irritant
(R)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene ≥95.0% (by GC)
Catalog # TCIAA2128-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %
  • Specific rotation [a]20/D:
    -100.0 to -112.0 deg (C=1, MeOH)

Specification Test Results

About this item

 :
 :150.22 g/mol
Flash Pt:74°C
 :GHS07
CAS Number:911200-98-1
Formula:C₁₀H₁₄O