


Acetonitrile-[D₃] ≥99.0% (by GC)
Catalog # TCIAA2018-10ML
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Degree of deuteration:min. 99.8 atom%D
Specification Test Results
About this item
:
:44.03 g/mol
Flash Pt:6°C
EINECS:218-616-5
UN:1648
ADR:3,II
:GHS02
:GHS06
:GHS08
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881