2-Acetamido-3-O-benzyl-4,6-O-benzylidene-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
TCIAA1813-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:-58.0 to -63.0 deg (C=0.5,pyridine)
- Solubility in Pyridine:very faint turbidity
:
:424.46 g/mol
:Freezer
:80887-27-0
:MFCD23104224
:C₂₂H₂₄N₄O₅
:MFCD06797052