Order Entry
Denmark
ContactUsLinkComponent
2-Acetamido-3-O-allyl-4,6-O-benzylidene-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
2-Acetamido-3-O-allyl-4,6-O-benzylidene-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
Catalog # TCIAA1812-1G
Supplier:  TCI
CAS Number:  
undefined
2-Acetamido-3-O-allyl-4,6-O-benzylidene-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
Catalog # TCIAA1812-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    -74.0 to -78.0 deg (C=0.5,pyridine)
  • Solubility in Pyridine:
    very faint turbidity

Specification Test Results

About this item

 :
 :374.40 g/mol
Storage Temperature:Freezer
Formula:C₁₈H₂₂N₄O₅
MDL Number:MFCD06797051