1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone ≥98.0% (by GC)
Catalog # TCIAA1629-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:82.0 to 86.0 deg-C
Specification Test Results
About this item
:
:178.19 g/mol
EINECS:220-726-3
CAS Number:2879-20-1
Formula:C₁₀H₁₀O₃
MDL Number:MFCD00006823