1-[(2S)-3-(Acetylthio)-2-methylpropionyl]-L-proline hydrate ≥90.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:64838-55-7
:
Formula:C₁₁H₁₇NO₄S
MDL Number:MFCD01860852
:259.32 g/mol
EINECS:265-250-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 90.0% (calcd.on anh.substance)
- Specific rotation [a]20/D:-155.0 to -165.0 deg (C=1, MeOH)
- Solubility in Methanol:very faint turbidity
- Water:max. 10.0 %
Specification Test Results
Frequently Bought Together