Ethyl-2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate ≥98.0% (by titrimetric analysis)
TCIAA1355-10G
: TCI
:
- Pk:10 g
- Seq:3236
- Appearance:White to Yellow to Green powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:114.0 to 117.0 deg-C
:
:225.31 g/mol
:224-823-1
:4506-71-2
:C₁₁H₁₅NO₂S
:MFCD00005862