Order Entry
Denmark
ContactUsLinkComponent
 
 
1H,1H,2H,2H-Heptadecafluorodecyl acrylate ≥97.0% (by GC) stabilised
  TCIAA1330-10G
 :  TCI
 :  
 
1H,1H,2H,2H-Heptadecafluorodecyl acrylate ≥97.0% (by GC) stabilised
  TCIAA1330-10G
 :  TCI
 :  

 

  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :518.17 g/mol
 :132°C
 :Stabilised with BHT + TBC
 :248-722-7
 :GHS07
 :27905-45-9
 :C₁₃H₇F₁₇O₂
 :MFCD00042306