Calcium alginate
Supplier: TCI
Certificates
About this item
Synonyms: Alginic acid calcium salt
CAS Number:9005-35-0
:
MDL Number:MFCD00143567
Merck Index:14,00242
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Drying loss:max. 20.0 %
- Calcium:7.0 to 10.0 % (calcd.on dried substance)
Specification Test Results
Related Information
Frequently Bought Together