Order Entry
Denmark
ContactUsLinkComponent
 
Monopotassium 7-amino-1,3-naphthalenedisulphonate Hydrate ≥98.0% (by HPLC)
 :  TCI
 
Monopotassium 7-amino-1,3-naphthalenedisulphonate Hydrate ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Drying loss:
    max. 5.0 %

 

Frequently Bought Together