


Trichloroacetyl isocyanate for NMR spectroscopy
:3019-71-4
:
:C₃Cl₃NO₂
:MFCD00002033
:188.4 g/mol
:80…85 °C (20 mmHg)
:1.59 g/cm³
:65 °C
:221-165-7
:2927
:6.1,II
:GHS06
:GHS08
:GHS05
:Refrigerator
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4799 to 1.4819 (20°C, 589 nm)
- GC:≥97 %
Frequently Bought Together