Order Entry
Denmark
ContactUsLinkComponent
 
Methylene blue trihydrate, Reagent Grade
Catalog # 0722-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Methylene blue trihydrate, Reagent Grade
Catalog # 0722-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bottle, amber
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Arsenic:
    <0.0008%
  • Copper:
    ≤0.03%
  • Dye Content (Dry Basis):
    98 – 103%
  • Em (661nm, Water):
    ≥69000
  • Identification:
    PASS/FAIL
  • Loss on Drying:
    ≤15.0%
  • Residue on Ignition:
    ≤1.0%
  • Zinc:
    ≤0.02%
  • Colour Index 52015:
    52015

Specification Test Results

About this item

Synonyms: 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride trihydrate, C.I.52015, Methylthioninium chloride trihydrate, Basic blue 9, Swiss Blue, Methylene blue
 :
 :373.9 g/mol
EINECS:200-515-2
 :GHS07
Storage Temperature:Ambient
CAS Number:7220-79-3
Formula:C₁₆H₁₈ClN₃S·3H₂O
MDL Number:MFCD00150008