Order Entry
Denmark
Orders LinkContactUsLinkComponent
L(-)-Cystine

L(-)-Cystine

Glass bottle for solids, 100 mg
Catalog # EDQMC3300000
Supplier:   EDQM
L(-)-Cystine
Catalog # EDQMC3300000
Supplier:  EDQM
CAS Number:  56-89-3
L(-)-Cystine

Glass bottle for solids, 100 mg

CAS Number:  56-89-3
Price
1.920,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
  • CAS Number:56-89-3
  • Formula:HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
  • MDL Number:MFCD00064228
  • Molecular Weight:240.3 g/mol
  • Merck Index:13,02811
  • Boiling Pt:428 °C (1013 hPa)
  • Melting Pt:261…262 °C
  • Density:1.677 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-296-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.