D-(-)-Serine 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (R)-(-)-Serine, H-D-Ser-OH
- CAS Number:312-84-5
- Formula:C₃H₇NO₃
- MDL Number:MFCD00004269
- Molecular Weight:105.09 g/mol
- Merck Index:12,08604
- Melting Pt:220 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-229-4
3 Options Available Below
Product Details & Documents
Synonyms: (R)-(-)-Serine, H-D-Ser-OH
- CAS Number:312-84-5
- Formula:C₃H₇NO₃
- MDL Number:MFCD00004269
- Molecular Weight:105.09 g/mol
- Merck Index:12,08604
- Melting Pt:220 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-229-4
Documents
ACRO22707
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 % (105°C, 3 h)
- Sulfated ash:≤0.1 %
- Specific optical rotation:-13.0° to -16.0° (20°C, 589 nm) (c=10, 2 N HCl)
- Titration with HClO4:97.5 to 102.5 %
- Other amino acids:≤0.5 % (TLC)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



