


Hexachloroplatinic(IV) acid hexahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2890-03
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Solubility in Alcohol:Passes Test
- Assay (as Pt):≥37.50 %
- Alkalies and Other Salts (as SO₄):≤0.05 %
- Suitability for Potassium Determination:Passes Test
Specification Test Results
About this item
:
:517.9 g/mol
Merck Index:13,07609
Melting Pt:150 °C
Density:2.434 g/cm³ (20 °C)
EINECS:241-010-7
UN:2507
ADR:8,III
:GHS06
:GHS05
:GHS08
CAS Number:18497-13-7
Formula:H₂PtCl₆·6H₂O
MDL Number:MFCD00149910