Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, Naphthalene 1,000 µg/ml in methanol
Organic reference standard, Naphthalene 1,000 µg/ml in methanol
Supplier:  Restek
Certificates
Organic reference standard, Naphthalene 1,000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    91-20-3
  • Formula:
    C₁₀H₈
  • Description:
    Naphthalene

Specifications

Frequently Bought Together