Order Entry
Denmark
ContactUsLinkComponent
Pesticide reference standard, 2,4-Dinitrophenol 1,000 µg/ml in methanol
Pesticide reference standard, 2,4-Dinitrophenol 1,000 µg/ml in methanol
Supplier:  Restek
Certificates
Pesticide reference standard, 2,4-Dinitrophenol 1,000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    51-28-5
  • Formula:
    C₆H₄N₂O₅
  • Description:
    2,4-Dinitrophenol

Specifications

Frequently Bought Together