

Potassium bromate concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Catalog # 88592.LQ
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
CAS Number:7758-01-2
:
Formula:KBrO₃
MDL Number:MFCD00011359
:167 g/mol
Storage Temperature:Ambient
EINECS:231-829-8
UN:3139
ADR:5.1,III
:GHS03
:GHS08