Order Entry
Denmark
ContactUsLinkComponent
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur., Sigma-Aldrich®
Corrosive
Irritant
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur., Sigma-Aldrich®
Catalog # 1.10789.0001
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur., Sigma-Aldrich®
Catalog # 1.10789.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    45 mL
  • Seq:
    2365

Specifications

About this item

 :
 :154.24 g/mol
ADR:8,
 :M-Clarity™ quality level = MQ200
EINECS:231-728-9
UN:1789
 :GHS05
 :GHS07
CAS Number:7705-07-9
Formula:Cl₃Ti
MDL Number:MFCD00011266