Order Entry
Denmark
ContactUsLinkComponent
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution
Certificates
undefined
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Solution
  • Titration Iodometric:
    10 to 15 %
  • Iron (Fe):
    ≤5 ppm
  • Free alkali (NaOH):
    ≤1.0 %

Specification Test Results

Related Information

Frequently Bought Together