Order Entry
Denmark
ContactUsLinkComponent
Sodium chloride 9 g/l in aqueous solution
Sodium chloride 9 g/l in aqueous solution
Catalog # 5929.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium chloride 9 g/l in aqueous solution
Catalog # 5929.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (W/V):
    0.891 to 0.909 %
  • Appearance:
    Clear colourless liquid
  • ZRAW:
    Passes test Ph. Eur.

Specification Test Results

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477