Order Entry
Denmark
ContactUsLinkComponent
Lactose monohydrate
Lactose monohydrate
Catalog # ROTH8921.5
Supplier:  Roth Carl
CAS Number:  
Lactose monohydrate
Catalog # ROTH8921.5
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Milk sugar, Lactobiose
 :
 :360.32 g/mol
Melting Pt:223
Storage Temperature:Ambient
EINECS:200-559-2
CAS Number:10039-26-6
Formula:C₁₂H₂₂O₁₁·1H₂O