Lactose monohydrate
Supplier: Roth Carl
Certificates
About this item
Synonyms: Milk sugar, Lactobiose
CAS Number:10039-26-6
:
Formula:C₁₂H₂₂O₁₁·1H₂O
Molecular Weight:360.32 g/mol
Melting Pt:223
Storage Temperature:Ambient
EINECS:200-559-2
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together