Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lubricants
VWR® TECHNICAL Vacuum oil for pump
VWR® TECHNICAL Vacuum oil for pump
9930.5000
:
VWR Chemicals
VWR® TECHNICAL Vacuum oil for pump
9930.5000
:
VWR Chemicals
Description:
VWR® TECHNICAL Vacuum oil for pump
CAS No.:
8012-95-1
Pack type:
Plastic container
Pk:
5 L