Order Entry
Denmark
ContactUsLinkComponent
Sodium chloride regeneration salt, TECHNICAL for water softening
  99062.4000
 :  
undefined
Sodium chloride regeneration salt, TECHNICAL for water softening
  99062.4000
 :  
  • Pack type:
    Plastic bucket
  • Pk:
    4 kg
  • Seq:
    4102
  • Environmentally Preferable:
  • Identification:
    Passes test
 : Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
 :1413 °C (1013 hPa)
 :801 °C
 :2.16 g/cm³ (20 °C)
 :Ambient
 :231-598-3
 :7647-14-5
 :NaCl
 :MFCD00003477