Order Entry
Denmark
ContactUsLinkComponent
Ethanol 70% ≥70% (v/v), GPR RECTAPUR®
Ethanol 70% ≥70% (v/v), GPR RECTAPUR®
Catalog # 98193.418
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ethanol 70% ≥70% (v/v), GPR RECTAPUR®
Catalog # 98193.418
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L
  • Seq:
    5214
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99 %
  • Assay (V/V):
    69.0-72.0 %
  • Evaporation residue:
    Max. 50 ppm

Specification Test Results

About this item

Synonyms: Ethyl alcohol
 :
 :46.07 g/mol
Boiling Pt:78.3
Flash Pt:22 °C
EINECS:200-578-6
UN:1170
 :GHS02
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568
Melting Pt:-117
Density:0.85 g/cm³ (20 °C)
ADR:3 II
Merck Index:12,03806