Order Entry
Denmark
ContactUsLinkComponent
Orthoboric acid 10 g/l in aqueous solution, VOLUSOL®, with indicator
Orthoboric acid 10 g/l in aqueous solution, VOLUSOL®, with indicator
Catalog # 5732.5000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Orthoboric acid 10 g/l in aqueous solution, VOLUSOL®, with indicator
Catalog # 5732.5000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay (W/V):
    0.9 - 1.1 %
  • Appearance:
    Clear green liquid
  • Sensitivity:
    Passes test

Specification Test Results

About this item

 :
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337