Order Entry
Denmark
ContactUsLinkComponent
Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP) 10 mM in buffer (Tris-HCl, pH 7.5)
Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP) 10 mM in buffer (Tris-HCl, pH 7.5)
Catalog # 40033.
Supplier:  Biotium
CAS Number:  
Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP) 10 mM in buffer (Tris-HCl, pH 7.5)
Catalog # 40033.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Plastic vial
  • Pk:
    25 µl
  • Seq:
    1166

Specifications

About this item

 :
 :902.5 g/mol
Storage Temperature:Freezer
Formula:C₂₈H₄₁N₆O₁₈P₃SLi₄