Murexide analytical reagent
Plastic bottle for solids, 10 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₈N₆O₆
- MDL Number:MFCD00012777
- Molecular Weight:284.19 g/mol
- Merck Index:13,06330
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-266-6
Documents
Certificates
Search by Lot NumberP20_VWRC25717.120_SPEC_EN.pdf spec_VWRC25717.120 spec_VWRC25717.120
Product specification
VWR® Indicators
Product Literature
Specifications
Specification Test Results
- Suited for complexometry indicator:Passes test
Specifications
- Pack type:Plastic bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



