Order Entry
Denmark
ContactUsLinkComponent
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98.5% analytical reagent
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98.5% analytical reagent
Catalog # 20308.156
Supplier:  VWR Chemicals
CAS Number:  
undefined
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98.5% analytical reagent
Catalog # 20308.156
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 98.5 %
  • Ignition residue (SO4):
    Max. 0.2 %
  • Water:
    Max. 2.0 %

Specification Test Results

About this item

 :
Merck Index:13,03560
Melting Pt:240…244 °C
 :380.35 g/mol
Storage Temperature:Ambient
EINECS:200-651-2
CAS Number:67-42-5
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291