Order Entry
Denmark
ContactUsLinkComponent
Apiezon® wax W, sticks
Apiezon® wax W, sticks
Catalog # 95060.195
Supplier:  M I MATERIALS
CAS Number:  
Apiezon® wax W, sticks
Catalog # 95060.195
Supplier:  M I MATERIALS
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Storage Temperature:Ambient