Order Entry
Denmark
ContactUsLinkComponent
 
VWR®, Casein acc. to Hammarsten
Catalog # 440203H
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Casein acc. to Hammarsten
Catalog # 440203H
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • CAS No.:
    9000-71-9
  • Protein/peptide name:
    Casein acc. to Hammarsten
  • Purity:
    Min 95 %

Specifications

About this item

High molecular weight phosphoprotein prepared from milk.

Assay (ex N, calc. on dried substance): Min 95%
Free acid (as lactic acid): Max 0,1%
Water-soluble matter: Max 0,2%
Fats: Max 0,1%
Sulfated ash (800 °C): Max 1,5%
Microbiological test: passes test
Lactose (HPLC): Max 0,5%
Glucose (HPLC): Max 0,5%