X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98.0%
Catalog # 437132J
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Assay:Min. 98.0 %
- Identification:Passes test
- Appearance:White crystalline powder
- Spec.opt. rotation (1 %;DMF/water 50/50):60.0 - 64.0 °
- Water:Max. 1 %
Specification Test Results
About this item
:
:408.63 g/mol
Melting Pt:237…239 °C
Storage Temperature:Freezer
EINECS:230-640-8
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666