Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® <i>Trichoderma viride</i> Cellulase

VWR® Trichoderma viride Cellulase

Glass bottle, 50 g
Catalog # 390744E
Supplier:   VWR Chemicals
VWR® <i>Trichoderma viride</i> Cellulase
Catalog # 390744E
Supplier:  VWR Chemicals
CAS Number:  
VWR® <i>Trichoderma viride</i> Cellulase
VWR® Trichoderma viride Cellulase

Glass bottle, 50 g

CAS Number:  
Price
3.680,00 kr
Log in for availability
1 * 50 g
Product Details & Documents

10,000 units is that quantity of enzyme which will degrade 2 cm² of cellulose filter paper (17 mg) in 1 minute in 0,1 M acetate buffer at pH 4,0.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Cat. No.:
    390744E
  • MDL No.:
    MFCD00081510
  • Species:
    Trichoderma viride
  • Enzyme name:
    Cellulase
  • CAS No.:
    9012-54-8

Specifications