Order Entry
Denmark
ContactUsLinkComponent
 
Biotin-11-CTP (Biotin-11-cytidine-5′-triphosphate, tetralithium salt) 10 mM in buffer (TRIS-HCl pH 7.5)
Catalog # 40035.
Supplier:  Biotium
CAS Number:  
Biotin-11-CTP (Biotin-11-cytidine-5′-triphosphate, tetralithium salt) 10 mM in buffer (TRIS-HCl pH 7.5)
Catalog # 40035.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Plastic vial
  • Pk:
    25 µl
  • Seq:
    1166

Specifications

About this item

 :
 :901.42 g/mol
Formula:C₂₈H₄₂N₇O₁₇P₃SLi₄