Order Entry
Denmark
ContactUsLinkComponent
Bromocresol green sodium salt, AnalaR NORMAPUR® ACS
Bromocresol green sodium salt, AnalaR NORMAPUR® ACS
Catalog # 85720.100
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bromocresol green sodium salt, AnalaR NORMAPUR® ACS
Catalog # 85720.100
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Identification (UV/VIS):
    Passes test
  • Clarity of solution (1 g/L in water):
    Passes test
  • Visual transition interval:
    pH 3,8 - 5,4
  • Loss on drying (110°C) :
    Max. 5,0%
  • Absorption maximum λ (buffer pH 3.8):
    438,00 - 443,00 nm
  • Absorption maximum λ (buffer pH 5.4):
    615,00 - 618,00 nm
  • Spec. absorptivity (λ1; 0.01 g/l; pH 3.8; dry):
    220,00 - 270,00
  • Spec. absorptivity (λ1; 0.01 g/l; pH 5.4; dry):
    460,00 - 570,00
  • Loss on drying (110°C):
    ≤ 5,00 %
  • Transition range (according to ACS) :
    Passes test

Specification Test Results

About this item

 :
Melting Pt:225 °C
 :720 g/mol
Storage Temperature:Ambient
EINECS:263-657-4
CAS Number:62625-32-5
 :67763-24-0
Formula:C₂₁H₁₃Br₄NaO₅S
MDL Number:MFCD00148898