Order Entry
Denmark
ContactUsLinkComponent
 
Boron trifluoride diethyl ether complex, GPR RECTAPUR® for synthesis
 
undefined
Boron trifluoride diethyl ether complex, GPR RECTAPUR® for synthesis
 

 

  • Assay (calculated as BF₃):
    45 - 50 %
  • Density (20/4):
    1,124 - 1,127
  • Refractive index (20 °C; 589 nm):
    1,343 - 1,344

 

Frequently Bought Together