Mouse Recombinant IL-22 (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Interleukin 22 (IL-22), also called IL-TIF, is an IL-10 family member that is produced by activated dendritic cells and T lymphocytes. IL-22 signals via a heteroduplex receptor consisting of IL-22R and IL-10RB chains. IL-22 is a potent mediator of cellular inflammatory responses and wound healing.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: Production of IL-10 from human COLO-205 cells, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Form:Lyophilised
- Gene ID:Q9JJY9
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:IL-TIF
- Protein/peptide name:IL-22
- Purity:≥95%
- Molecular weight:16,8 kDa
- Sequence:MLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACV
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together