N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane (99:1) for GC derivatisation
Supplier: TCI
Certificates
About this item
Synonyms: BSTFA+TMCS (99:1)
CAS Number:25561-30-2
:
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269
Boiling Pt:142 °C (lit.)
Molecular Weight:257.40 g/mol
Melting Pt:–41 °C (lit.)
Flash Pt:7 °C (lit.)
UN:2924
ADR:3 II
:GHS02
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- High boiling impurity of Silylation agent:to pass test
- Suitability for Melamine-analysis:to pass test
Specification Test Results
Frequently Bought Together