Human Recombinant CNTF (from E. coli)
Supplier: Shenandoah Biotechnology
About this item
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis.CNTF also plays an important protective role during nervous system injury.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
5 Options Available Below
Product Details & Documents
Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that promotes the survival of neuronal cell populations, neurite outgrowth, and neurotransmitter synthesis.CNTF also plays an important protective role during nervous system injury.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- SHBT100-59AF-5UG
- SHBT100-59AF-20UG
- SHBT100-59AF-100UG
- SHBT100-59AF-500UG
- SHBT100-59AF-1MG
- Protein/peptide name:
- CNTF (AF)
- CNTF (AF)
- CNTF (AF)
- CNTF (AF)
- CNTF (AF)
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P26441
- P26441
- P26441
- P26441
- P26441
- Sequence:
- MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
- MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
- MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
- MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
- MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- Y
- Y
- Y
- Y
- Y
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:
- Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mg
- Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mg
- Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mg
- Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mg
- Bioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM Sodium phosphate, pH 7,5
- 10 mM Sodium phosphate, pH 7,5
- 10 mM Sodium phosphate, pH 7,5
- 10 mM Sodium phosphate, pH 7,5
- 10 mM Sodium phosphate, pH 7,5
- Molecular weight:
- 22,9 kDa
- 22,9 kDa
- 22,9 kDa
- 22,9 kDa
- 22,9 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.SHBT100-59AF-5UGProtein/peptide nameCNTF (AF)Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP26441SequenceMAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKMProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7,5Molecular weight22,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-59AF-20UGProtein/peptide nameCNTF (AF)Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP26441SequenceMAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKMProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7,5Molecular weight22,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-59AF-100UGProtein/peptide nameCNTF (AF)Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP26441SequenceMAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKMProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7,5Molecular weight22,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-59AF-500UGProtein/peptide nameCNTF (AF)Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP26441SequenceMAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKMProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7,5Molecular weight22,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Specifications:
More
Specifications
Cat. No.SHBT100-59AF-1MGProtein/peptide nameCNTF (AF)Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP26441SequenceMAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKMProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0,1Biological activityBioactivity Assay: TF-1 cell proliferation, ED₅₀ ≤325 ng/ml, Bioactivity ≥3,1×10³ units/mgPurity≥95%GradeRUOFormulation10 mM Sodium phosphate, pH 7,5Molecular weight22,9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay



