Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant GDNF (from E. coli)
Human Recombinant GDNF (from E. coli)
Certificates
undefined
Human Recombinant GDNF (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity Assay: C₆ proliferation, ED₅₀ ≤3 µg/ml, Bioactivity ≥3,3×10² units/mg
  • Gene ID:
    P39905
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    ATF-1
  • Protein/peptide name:
    GDNF (AF)
  • Purity:
    ≥95%
  • Molecular weight:
    15,2/30,4 kDa
  • Sequence:
    MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium citrate, 100 mM Sodium chloride, pH 4,0
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together