Order Entry
Denmark
ContactUsLinkComponent
TMS-HT (=HMDS and TMCS in anhydrous pyridine) for gas chromatography
Flammable
Corrosive
Irritant
Health
Environment
TMS-HT (=HMDS and TMCS in anhydrous pyridine) for gas chromatography
Catalog # TCIAA5604-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Environment
TMS-HT (=HMDS and TMCS in anhydrous pyridine) for gas chromatography
Catalog # TCIAA5604-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
  • Effect test of TMS derivatization:
    Effective as derivatization agent Glucose
  • High boiling impurity of Silylation agent:
    No impurity higher than Isopropyl myristate(50 ppm) after 5 min + hold up time

Specification Test Results

About this item

 :
Flash Pt:20 °C
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
 :GHS09
Storage Temperature:Refrigerator
MDL Number:MFCD00192427