Sodium 3-(Trimethylsilyl)-1-propanesulfonate, 1H NMR Standard for D₂O Solvent
: TCI
Suitable for NMR-analysis.
- CAS No.:2039-96-5
- Formula:C₆H₁₅NaO₃Ssi
- Description:Sodium 3-(Trimethylsilyl)-1-propanesulfonate
- Appearance:White to Almost white powder to crystal
Frequently Bought Together