Order Entry
Denmark
ContactUsLinkComponent
p-Xylene, tag closed flash point check fluid
p-Xylene, tag closed flash point check fluid
Catalog # TCIAX0044-250ML
Supplier:  TCI
CAS Number:  
undefined
p-Xylene, tag closed flash point check fluid
Catalog # TCIAX0044-250ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • CAS No.:
    106-42-3
  • Description:
    p-Xylene, Purity (GC): min. 99.0 %
  • Formula:
    C₆H₄(CH₃)₂
  • Description:
    p-Xylene
  • Flash point:
    "27 °C"
  • Boiling point:
    139 °C
  • Appearance:
    Colorless clear liquid

Specifications

About this item